Boc-L-tyrosine methyl ester 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-Tyr-OMe, N-Boc-L-tyrosine Methyl Ester

44202-0050 44202-0250
TS44202-0050EA 70 USD
TS44202-0050 TS44202-0250
Boc-L-tyrosine methyl ester 97%
Boc-L-tyrosine methyl ester
Formula: C₁₅H₂₁NO₅
MW: 295.3 g/mol
Melting Pt: 106…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00191181
CAS Number: 4326-36-7

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Specific optical rotation +48° to +52° (20°C, 589 nm) (c=1 in CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR