L-Valine methyl ester hydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Val-OMe.HCl, L-2-Aminoisovaleric Acid Methyl Ester Hydrochloride, L-Methyl valinate hydrochloride, H-Val-OMe·HCl

22926-0100 22926-0500
TS22926-0100EA 56 USD
TS22926-0100 TS22926-0500
L-Valine methyl ester hydrochloride 99%
L-Valine methyl ester hydrochloride
Formula: C₆H₁₄ClNO₂
MW: 167.63 g/mol
Melting Pt: 171…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00012497
CAS Number: 6306-52-1

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Solubility (5 % in water) Clear colorless to light yellow
Specific optical rotation +14° to +17° (20°C, 589 nm) (c=2, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR