N-Cbz-L-serine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Carbobenzyloxy-L-serine, Z-Ser-OH, N-Cbz-L-serine

22951-0050 22951-0250
TS22951-0050EA 31 USD
TS22951-0050 TS22951-0250
N-Cbz-L-serine 99+%
N-Cbz-L-serine
Formula: C₁₁H₁₃NO₅
MW: 239.2 g/mol
Melting Pt: 115…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00002662
CAS Number: 1145-80-8

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 115 to 120 °C
Water ≤0.5 % (K.F.)
HPLC ≥99.0 %
Specific optical rotation +5.5° to +6.5° (20 °C, 589 nm) (c=7, AcOH)
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR