2-(4-Isobutylphenyl)propanoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(±)-Ibuprofen, [(±)-2-(4-Isobutylphenyl)-propionic Acid, Ibuprofen
25861-0100
25861-0010
25861-0500
TS25861-0100EA
243
USD
TS25861-0100
TS25861-0010
TS25861-0500
2-(4-Isobutylphenyl)propanoic acid 99%
2-(4-Isobutylphenyl)propanoic acid
Formula:
C₁₃H₁₈O₂ MW: 206.3 g/mol Boiling Pt: 157 °C (4 mmHg) Melting Pt: 75…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010393 CAS Number: 15687-27-1 |
Specification Test Results
Appearance | White to almost white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 75 to 78 °C |
Titration with NaOH | 98.5 to 101.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...