Boc-D-Alanine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Boc-D-alanine, Boc-D-Ala-OH

27572-0050 27572-0250
TS27572-0050EA 50 USD
TS27572-0050 TS27572-0250
Boc-D-Alanine 99+%
Boc-D-Alanine
Formula: C₈H₁₅NO₄
MW: 189.2 g/mol
Melting Pt: 81…84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063123
CAS Number: 7764-95-6

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 81 to 84 °C
HPLC ≥99.0 % (ELSD)
Specific optical rotation +25° to +27° (20 °C, 589 nm) (c=2, CH3COOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR