N-Boc-L-serine, anhydrous 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid, Boc-Ser-OH, N-(tert-Butoxycarbonyl)-L-serine, Boc-L-serine, N-(tert-Butoxycarbonyl)-L-serine hydrate, BOC-L-Serine hydrate

44256-0050 44256-0250
TS44256-0050EA 52 USD
TS44256-0050 TS44256-0250
N-Boc-L-serine, anhydrous 99%
N-Boc-L-serine
Formula: C₈H₁₅NO₅
MW: 205.21 g/mol
Melting Pt: 84…89 °C
Flash Pt: 186 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037243
CAS Number: 3262-72-4

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥98.5 %
Specific optical rotation -6.5° to -8.5° (20°C, 589 nm) (c=2 in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR