N-Boc-L-serine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid, Boc-Ser-OH, N-(tert-Butoxycarbonyl)-L-serine, Boc-L-serine, N-(tert-Butoxycarbonyl)-L-serine hydrate, BOC-L-Serine hydrate

27566-0050 27566-0250 27566-1000
TS27566-0050EA 45 USD
TS27566-0050 TS27566-0250 TS27566-1000
N-Boc-L-serine 98%
N-Boc-L-serine
Formula: C₈H₁₅NO₅
MW: 205.21 g/mol
Melting Pt: 46…52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037243
CAS Number: 3262-72-4

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 46 to 52 °C
Water 7.5 to 8.0 % (K.F.)
Specific optical rotation -7° to -8° (20 °C, 589 nm) (c=2, H2O)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR