(S)-Methyl-2-aminopropanoate hydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Ala-OMe·HCl, H-Ala-OMe.HCl, L-Alanine methyl ester hydrochloride, (S)-2-Aminopropionic Acid Methyl Ester Hydrochloride

30836-0050 30836-0250
TS30836-0050EA 29 USD
TS30836-0050 TS30836-0250
(S)-Methyl-2-aminopropanoate hydrochloride 99%
(S)-Methyl-2-aminopropanoate hydrochloride
Formula: C₄H₉NO₂·HCl
MW: 139.6 g/mol
Melting Pt: 109…111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063663
CAS Number: 2491-20-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Specific optical rotation +6° to +8° (24°C, 589 nm) (c=1.6, CH3OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR