N-Carbobenzyloxy-L-valine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Cbz-L-valine, Z-L-Val-OH, Z-Val-OH

27594-0050 27594-0250
TS27594-0050EA 27 USD
TS27594-0050 TS27594-0250
N-Carbobenzyloxy-L-valine 99+%
N-Carbobenzyloxy-L-valine
Formula: C₁₃H₁₇NO₄
MW: 251.3 g/mol
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008922
CAS Number: 1149-26-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.2 %
HPLC ≥98.0 %
Specific optical rotation -3.2° to -4.8° (20°C, 589 nm) (c=2, acetic acid)
Titration with NaOH ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR