N-Carbobenzyloxy-L-alanine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Cbz-L-alanine, Z-Ala-OH, (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid, N-Benzyloxycarbonyl-L-alanine, ((Benzyloxy)carbonyl)-L-alanine

16900-0050 16900-0250 16900-1000
TS16900-0050EA 25 USD
TS16900-0050 TS16900-0250 TS16900-1000
N-Carbobenzyloxy-L-alanine 98%
N-Carbobenzyloxy-L-alanine
Formula: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00002640
CAS Number: 1142-20-7

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
TLC ≥97.5 %
Appearance of solution (5% soln. in MeOH) clear, colorless
Specific optical rotation -14° to -16° (24°C, 589 nm) (c=2, AcOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR