Methyl-2-amino-3-hydroxypropanoate hydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl DL-serinate hydrochloride, H-DL-Ser-OMe·HCl, 3-hydroxy-1-methoxy-1-oxopropan-2-aminium chloride, methyl 2-amino-3-hydroxypropanoate, DL-Serine Methyl Ester Hydrochloride, H-DL-Ser-OMe.HCl

20745-0100 20745-1000
TS20745-0100EA 40 USD
TS20745-0100 TS20745-1000
Methyl-2-amino-3-hydroxypropanoate hydrochloride 99%
Methyl-2-amino-3-hydroxypropanoate hydrochloride
Formula: C₄H₁₀ClNO₃
MW: 155.578 g/mol
Melting Pt: 130…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012593
CAS Number: 5619-04-5

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR