(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride 98%

Supplier: Thermo Scientific Chemicals

13032-0100 13032-0500
TS13032-0100EA 46 USD
TS13032-0100 TS13032-0500
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride 98%
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride
Formula: C₁₀H₁₄ClNO₂
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00012489
CAS Number: 7524-50-7

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 156 to 160 °C
Titration Argentometric ≥97.5 %
Specific optical rotation +37° to +39° (20 °C, 589 nm) (c=2, C2H5OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR