N-Boc-L-alanine 99+%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Alpha-t-Boc-L-alanine, (S)-2-((tert-Butoxycarbonyl)amino)propanoic acid, BOC-L-Alanine, Boc-Ala-OH
27549-0100
27549-0500
TS27549-0100EA
44
USD
TS27549-0100
TS27549-0500
N-Boc-L-alanine 99+%
N-Boc-L-alanine
Formula:
C₈H₁₅NO₄ MW: 189.21 g/mol Melting Pt: 81…84 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037225 CAS Number: 15761-38-3 |
Specification Test Results
Appearance | White to off-white Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 81 to 85 °C |
HPLC | ≥99.0 % (ELSD) |
Specific optical rotation | -25° to -27° (20 °C, 589 nm) (c=2, CH3COOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...