Boc-N-methyl-D-alanine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-N-Me-Ala-OH, N-Boc-N-methyl-L-alanine

44293-0010 44293-0050
TS44293-0010EA 78 USD
TS44293-0010 TS44293-0050
Boc-N-methyl-D-alanine 99%
Boc-N-methyl-D-alanine
Formula: C₉H₁₇NO₄
MW: 203.2 g/mol
Melting Pt: 88…92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037242
CAS Number: 16948-16-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Specific optical rotation -29° to -31° (20°C, 589 nm) (c=1 in EtOH)
Titration with NaOH 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR