(R)-1-Boc-piperidine-2-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-(+)-N-BOC-2-Piperidinecarboxylic acid, Boc-D-Homopro-OH, Boc-D-HoPro-OH, (R)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid

39925-0010 39925-0050
TS39925-0010EA 46 USD
TS39925-0010 TS39925-0050
(R)-1-Boc-piperidine-2-carboxylic acid 98%
(R)-1-Boc-piperidine-2-carboxylic acid
Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00237380
CAS Number: 28697-17-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 120 to 125 °C
Specific optical rotation +62.0° to +65.0° (20 °C, 589 nm) (c=1%, acetic acid)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR