2-(1H-Imidazol-5-yl)acetic acid hydrochloride 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Imidazoleacetic acid hydrochloride, Imidazole-4-acetic acid monohydrochloride
20496-0010
20496-0050
TS20496-0010EA
124
USD
TS20496-0010
TS20496-0050
2-(1H-Imidazol-5-yl)acetic acid hydrochloride 99%
2-(1H-Imidazol-5-yl)acetic acid hydrochloride
Formula:
C₅H₇ClN₂O₂ MW: 162.58 g/mol Melting Pt: 218…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012698 CAS Number: 3251-69-2 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 221 to 225 °C |
Titration Argentometric | ≥98.5 % |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...