(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-Homopro-OH, 1-(tert-Butoxycarbonyl)-L-pipecolic Acid, Boc-HomoPro-OH, 1-Boc-L-pipecolic Acid, (S)-1-Boc-2-piperidinecarboxylic Acid, Boc-Pip-OH, (S)-N-BOC-Piperidine-2-carboxylic acid

39783-0050 39783-0250
TS39783-0050EA 131 USD
TS39783-0050 TS39783-0250
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid 98%
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid
Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00151904
CAS Number: 26250-84-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Specific optical rotation -62° to -65° (20°C, 589 nm) (C=1, acetic acid)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR