(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Cbz-Phe-OH, N-Cbz-L-phenylalanine, N-Carbobenzyloxy-L-phenylalanine, Z-Phe-OH

27596-0050 27596-0250
TS27596-0050EA 25 USD
TS27596-0050 TS27596-0250
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid 99+%
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid
Formula: C₁₇H₁₇NO₄
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00020418
CAS Number: 1161-13-3

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 84 to 88 °C
Water ≤0.2 % (K.F.)
HPLC ≥98.0 %
Specific optical rotation +4.5° to +5.5° (20 °C, 589 nm) (c=5, CH3COOH)
Titration with NaOH 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR