Acyclovir 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,9-dihydro-2-amino-9-((2-hydroxyethoxy)methyl)-6h-purin-6-on, Acycloguanosine, 2-amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6h-purin-6-one, Acyclovir, 9-[(2-Hydroxyethoxy)methyl]guanine, 2-Amino-1,9-dihydro-9-((2-hydroxyethoxy)methyl)-6H-purin-6-one, 2-Amino-1,9-dihydro-9-[(2-hydroxyethoxy)methyl]-6H-purin-6-one, Aciclovir, 2-Amino-9-((2-hydroxyethoxy)methyl)-1H-purin-6(9H)-one

44524-0100 44524-0500
TS44524-0100EA 40 USD
TS44524-0100 TS44524-0500
Acyclovir 98%
Acyclovir
Formula: C₈H₁₁N₅O₃
MW: 225.21 g/mol
Melting Pt: 256…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00057880
CAS Number: 59277-89-3

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR