1,3-Di-boc-2-(trifluoromethylsulfonyl)guanidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Bis(tert-butoxycarbonyl)-2-(trifluoromethanesulphonyl)guanidine, Goodman's Reagent, 1,3-Bis(tert-butoxycarbonyl)-2-(trifluoromethylsulphonyl)guanidine, 1,3-Di-Boc-2-(trifluoromethylsulphonyl)guanidine, 1,3-Di-Boc-2-(trifluoromethanesulphonyl)guanidine

39975-0010 39975-0050
TS39975-0010EA 156 USD
TS39975-0010 TS39975-0050
1,3-Di-boc-2-(trifluoromethylsulfonyl)guanidine 98%
1,3-Di-boc-2-(trifluoromethylsulfonyl)guanidine
Formula: C₁₂H₂₀F₃N₃O₆S
MW: 391.36 g/mol
Melting Pt: 111…113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683528
CAS Number: 207857-15-6

Order Now

Specification Test Results

Appearance White Solid
Infrared spectrum Conforms
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR