4-Iodo-1-tritylimidazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Iodo-1-trityl-1H-imidazole

45920-0050 459200010
TS45920-0050EA 224 USD
TS45920-0050 TS459200010
4-Iodo-1-tritylimidazole 98%
4-Iodo-1-tritylimidazole
Formula: C₂₂H₁₇IN₂
MW: 436.2900 g/mol
Melting Pt: 223…227.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179542
CAS Number: 96797-15-8

Order Now

Specification Test Results

Appearance White to off-white Powder or solid
Infrared spectrum Conforms
Melting point 222 to 229 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR