2,2-Diphenylglycine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: alpha,alpha-Diphenylglycine, Diphenylglycin, 2-Amino-2,2-diphenylacetic acid

17172-0050 17172-0250 17172-1000
TS17172-0050EA 84 USD
TS17172-0050 TS17172-0250 TS17172-1000
2,2-Diphenylglycine 98%
2,2-Diphenylglycine
Formula: C₁₄H₁₃NO₂
MW: 227.26 g/mol
Melting Pt: 244…247 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008048
CAS Number: 3060-50-2
EINECS: 221-299-6

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with TBAH 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR