N-Boc-L-tyrosine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: BOC-L-Tyrosine, Boc-Tyr-OH

27570-0050 27570-0250 27570-1000
TS27570-0050EA 47 USD
TS27570-0050 TS27570-0250 TS27570-1000
N-Boc-L-tyrosine 99+%
N-Boc-L-tyrosine
Formula: C₁₄H₁₉NO₅
MW: 281.3 g/mol
Melting Pt: 135…140 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00037179
CAS Number: 3978-80-1
EINECS: 223-613-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
HPLC ≥99.0 %
Specific optical rotation +36.5° to +39.5° (24°C, 589 nm) (c=1, dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR