N-Boc-DL-valine 98%

Supplier: Thermo Scientific Chemicals

42578-0100 42578-0500
TS42578-0100EA 79 USD
TS42578-0100 TS42578-0500
N-Boc-DL-valine 98%
N-Boc-DL-valine
Formula: C₁₀H₁₉NO₄
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00069973
CAS Number: 54895-12-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
NMR consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR