N(α)-Boc-L-tryptophan 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-Trp-OH , Boc-Trp-OH

27569-0050 27569-0250 27569-1000
TS27569-0050EA 48 USD
TS27569-0050 TS27569-0250 TS27569-1000
N(α)-Boc-L-tryptophan 97%
N(α)-Boc-L-tryptophan
Formula: C₁₆H₂₀N₂O₄
MW: 304.3 g/mol
Melting Pt: 134…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00065595
CAS Number: 13139-14-5
EINECS: 236-072-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1.0 % (K.F.)
HPLC ≥96.0 %
Specific optical rotation -19° to -23° (20°C, 589 nm) (c=1, CH3COOH)
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR