(±)-Boc-α-phosphonoglycine trimethyl ester 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyl 2-((tert-butoxycarbonyl)amino)-2-(dimethoxyphosphoryl)acetate
43921-0010
43921-0050
TS43921-0010EA
58
USD
TS43921-0010
TS43921-0050
(±)-Boc-α-phosphonoglycine trimethyl ester 97%
(±)-Boc-α-phosphonoglycine trimethyl ester
Formula:
C₁₀H₂₀NO₇P MW: 297.24 g/mol Melting Pt: 69.7…71.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD02178893 CAS Number: 89524-98-1 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...