1-Boc-piperidine-4-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acid, N-BOC-4-Piperidinecarboxylic acid, Boc-Inp-OH, 1-Boc-isonipecotic acid

38487-0010 38487-0050
TS38487-0010EA 41 USD
TS38487-0010 TS38487-0050
1-Boc-piperidine-4-carboxylic acid 98%
1-Boc-piperidine-4-carboxylic acid
Formula: C₁₁H₁₉NO₄
Melting Pt: 149…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00076999
CAS Number: 84358-13-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 149 to 153 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR