1-Boc-isonipecotic acid ethyl ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl N-BOC-4-piperidinecarboxylate, Ethyl N-Boc-piperidine-4-carboxylate

43150-0050 43150-0250
TS43150-0050EA 94 USD
TS43150-0050 TS43150-0250
1-Boc-isonipecotic acid ethyl ester 97%
1-Boc-isonipecotic acid ethyl ester
Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
Boiling Pt: 120…135 °C (.5 mmHg)
Density: 1.046 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01763998
CAS Number: 142851-03-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR