Coconut oil, pure, refined

Supplier: Thermo Scientific Chemicals

36547-1000 36547-5000
TS36547-1000EA 31 USD
TS36547-1000 TS36547-5000
Coconut oil, pure, refined
Coconut oil
Melting Pt: 23…26 °C
Flash Pt: >200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130817
CAS Number: 8001-31-8
EINECS: 232-282-8

Order Now

Specification Test Results

Appearance Colorless to white to light yellow Oily liquid or solid
Acid value ≤0.5 mg KOH/g
Iodine value 7 to 12 g I2/100 g
Peroxide value ≤5 meq O2/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR