Estradiol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
beta-Estradiol, 17β-Estradiol
43632-0050
43632-0250
TS43632-0050EA
336
USD
TS43632-0050
TS43632-0250
Estradiol 98%
Estradiol
Formula:
C₁₈H₂₄O₂ MW: 272.39 g/mol Melting Pt: 176…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003693 CAS Number: 50-28-2 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 173 to 179 °C |
Water | ≤3.5 % |
HPLC | ≥97.5 % |
Specific optical rotation | +76° to +83° (20 °C, 589 nm) (c=1, dioxane) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...