L-(-)-Fucose 97%

Supplier: Thermo Scientific Chemicals

22588-0250 22588-0010 22588-0050
TS22588-0250EA 682 USD
TS22588-0250 TS22588-0010 TS22588-0050
L-(-)-Fucose 97%
L(-)-Fucose
Formula: C₆H₁₂O₅
Melting Pt: 128…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00069812
CAS Number: 2438-80-4
EINECS: 219-452-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
HPLC ≥96.0 % (ELSD)
Specific optical rotation -72° to -78° (20°C, 589 nm) (c=4, H2O, NH3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR