Piceatannol

Supplier: Thermo Scientific Chemicals

Synonyms: (E)-4-(3,5-Dihydroxystyryl)benzene-1,2-diol

32845-2500 328450010
TS32845-2500EA 233 USD
TS32845-2500 TS328450010
Piceatannol
Piceatannol
Formula: C₁₄H₁₂O₄
Melting Pt: 235 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00221715
CAS Number: 10083-24-6

Order Now

Specification Test Results

Appearance Yellow to brown-pink Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR