N(α)-Boc-L-histidine 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: Boc-His-OH

27559-0010 27559-0050 27559-0250
TS27559-0010EA 32 USD
TS27559-0010 TS27559-0050 TS27559-0250
N(α)-Boc-L-histidine 99+%
N(α)-Boc-L-histidine
Formula: C₁₁H₁₇N₃O₄
MW: 255.3 g/mol
Melting Pt: 196…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00065576
CAS Number: 17791-52-5
EINECS: 241-768-9

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals and/or chunks
Infrared spectrum Conforms
TLC ≥99.0 %
Specific optical rotation +24.5° to +26.5° (20°C, 589 nm) (c=1, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR