N-Phenylglycine 93%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Anilinoacetic acid, 2-(Phenylamino)acetic acid

27608-0250 27608-1000
TS27608-0250EA 50 USD
TS27608-0250 TS27608-1000
N-Phenylglycine 93%
N-Phenylglycine
Formula: C₈H₉NO₂
Melting Pt: 121…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00014009
CAS Number: 103-01-5
EINECS: 203-070-2

Order Now

Specification Test Results

Appearance Ochre to yellow-brown Powder
Infrared spectrum Conforms
Melting point 121 to 127 °C
HPLC ≥91 %
Titration with TBAH ≥91 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR