Glycine hydrochloride (H-Gly-OH.HCl) 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Gly-OH·HCl, Glycine hydrochloride
41101-0025
41101-1000
41101-5000
TS41101-0025EA
682
USD
TS41101-0025
TS41101-1000
TS41101-5000
Glycine hydrochloride (H-Gly-OH.HCl) 98%
Glycine hydrochloride (H-Gly-OH.HCl)
Formula:
C₂H₆ClNO₂ MW: 111.53 g/mol Melting Pt: 173…177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012872 CAS Number: 6000-43-7 EINECS: 227-841-8 |
Specification Test Results
Appearance | White to almost white Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 175 to 180 °C |
Titration Argentometric | 97.5 to 102.5 % |
Water | ≤1.0 % (K.F.) |
Anticaking agent | 0.5 % (SiO2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...