N-Boc-glycine (boc-gly-OH) 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Boc-Gly-OH

15880-0100 15880-1000
TS15880-0100EA 58 USD
TS15880-0100 TS15880-1000
N-Boc-glycine (boc-gly-OH) 99+%
N-Boc-glycine (Boc-Gly-OH)
Formula: C₇H₁₃NO₄
MW: 175.2 g/mol
Melting Pt: 87…90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002690
CAS Number: 4530-20-5
EINECS: 224-864-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 85 to 90 °C
Water ≤0.5 % (K.F.)
HPLC ≥99.0 % (ELSD)
Titration with NaOH ≥99.0 %
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR