DL-Glutamic acid monohydrate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: DL-Glutamic Acid Monohydrate

11993-0010 11993-0250 11993-1000
TS11993-0010EA 388 USD
TS11993-0010 TS11993-0250 TS11993-1000
DL-Glutamic acid monohydrate 98+%
DL-Glutamic acid monohydrate
Formula: C₅H₉NO₄·1H₂O
MW: 165.15 g/mol (147.13 anhy)
Melting Pt: 194 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150703
CAS Number: 19285-83-7
EINECS: 210-522-2

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Water ≤15.5 % (K.F.)
Titration with NaOH ≥98.0 %
Specific optical rotation +31° to +35° (20°C, 589 nm) (c=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR