D-(-)-α-(4-Hydroxyphenyl)glycine 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: D(-)-4-Hydroxyphenylglycine, H-D-Phg(4-OH)-OH, (R)-(-)-2-(4-Hydroxyphenyl)glycine, (-)-4-Hydroxy-D-phenylglycine, D-(-)-p-Hydroxyphenylglycine

20132-0250 20132-1000
TS20132-0250EA 42 USD
TS20132-0250 TS20132-1000
D-(-)-α-(4-Hydroxyphenyl)glycine 98+%
D(-)-α-(4-Hydroxyphenyl)glycine
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004262
CAS Number: 22818-40-2
EINECS: 245-247-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Specific optical rotation -155° to -161° (20°C, 589 nm) (c=1, 1N HCl)
Titration with HClO4 ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR