L-Glutamic acid 5-benzyl ester 99%

Supplier: Thermo Scientific Chemicals

Synonyms: γ-Benzyl-L-Glutamate, 5-Benzyl L-Glutamate, gamma-Benzyl L-glutamate, H-Glu(OBzl)-OH

22567-0010 22567-0050 22567-0250
TS22567-0010EA 32 USD
TS22567-0010 TS22567-0050 TS22567-0250
L-Glutamic acid 5-benzyl ester 99%
L-Glutamic acid 5-benzyl ester
Formula: C₁₂H₁₅NO₄
Melting Pt: 162…167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002633
CAS Number: 1676-73-9
EINECS: 216-826-1

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation +26° to +29° (20°C, 589 nm) (c=2, 1N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR