Fmoc-D-phenylalanine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Fmoc-D-Phe-OH, N-Fmoc-D-phenylalanine

44171-0010 44171-0050
TS44171-0010EA 23 USD
TS44171-0010 TS44171-0050
Fmoc-D-phenylalanine 98%
Fmoc-D-phenylalanine
Formula: C₂₄H₂₁NO₄
MW: 387.4 g/mol
Melting Pt: 184…186.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00062955
CAS Number: 86123-10-6

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation +38° to +42° (20°C, 589 nm) (C=1 IN DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR