Ammonium 1-pyrrolidinecarbodithioate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyrrolidinedithiocarbamate ammonium salt, 1-Pyrrolidinecarbodithioic acid, ammonium salt, Ammonium pyrrolidine-1-carbodithioate, Pyrrolidine-1-Dithiocarboxylic Acid Ammonium Salt, APCD

16205-1000 16205-0250 16205-5000
TS16205-1000EA 178 USD
TS16205-1000 TS16205-0250 TS16205-5000
Ammonium 1-pyrrolidinecarbodithioate 98%
Ammonium 1-pyrrolidinecarbodithioate
Formula: C₅H₁₂N₂S₂
MW: 146.3 · 18.0 g/mol
Melting Pt: 150…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012720
CAS Number: 5108-96-3
EINECS: 225-834-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR