N-Acetyl-L-leucine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Ac-Leu-OH, Ac-Leu-OH

34701-0050 34701-0250
TS34701-0050EA 42 USD
TS34701-0050 TS34701-0250
N-Acetyl-L-leucine 99%
N-Acetyl-L-leucine
Formula: C₈H₁₅NO₃
MW: 173.2 g/mol
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00065131
CAS Number: 1188-21-2
EINECS: 214-706-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Solubility (5 % in ethanol) Clear colorless
Specific optical rotation -23° to -26° (20°C, 589 nm) (c=4, MeOH)
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR