Nalidixic acid sodium salt
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate
46355-0010
463550050
463550250
TS46355-0010EA
31
USD
TS46355-0010
TS463550050
TS463550250
Nalidixic acid sodium salt
Nalidixic acid sodium salt
Formula:
C₁₂H₁₁N₂NaO₃ MW: 254.22 g/mol Melting Pt: >280 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064376 CAS Number: 3374-05-8 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Water | ≤7 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...