Indomethacin

Supplier: Thermo Scientific Chemicals
Danger

45803-0050 45803-0250 45803-1000
TS45803-0050EA 41 USD
TS45803-0050 TS45803-0250 TS45803-1000
Indomethacin
Indomethacin
Formula: C₁₉H₁₆ClNO₄
MW: 357.80 g/mol
Melting Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00057095
CAS Number: 53-86-1
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline powder (May have a pink hue)
Infrared spectrum Conforms
Melting point 156 to 165 °C
Loss on drying ≤0.5 % (100 °C, 2 h, 5 mm Hg)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR