Iminodiacetic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
20497-0025
20497-0250
20497-5000
TS20497-0025EA
242
USD
TS20497-0025
TS20497-0250
TS20497-5000
Iminodiacetic acid 98+%
Iminodiacetic acid
Formula:
C₄H₇NO₄ Melting Pt: 243 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004280 CAS Number: 142-73-4 EINECS: 205-555-4 |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...