5-Bromo-2'-deoxyuridine (broxuridine) 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
22859-0010
22859-0025
22859-0100
22859-5000
TS22859-0010EA
127
USD
TS22859-0010
TS22859-0025
TS22859-0100
TS22859-5000
5-Bromo-2'-deoxyuridine (broxuridine) 99+%
5-Bromo-2'-deoxyuridine (broxuridine)
Formula:
C₉H₁₁BrN₂O₅ Melting Pt: 191…194 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006529 CAS Number: 59-14-3 EINECS: 200-415-9 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Water | ≤2 % (K.F.) |
HPLC | ≥99.0 % |
Ratio of Absorbances | 1.45 to 1.55 (A290/A260) |
Microbial contamination | ≤100 CFU/g (Total count) |
Bacterial endotoxins | ≤0.1 EU/mg (LAL-test) |
Related substances | ≤1.0 % (HPLC) Total impurities |
Specific optical rotation | +20° to +24° (20°C, 589 nm) (c=1, H2O) |
Heavy metals (as Pb) | ≤10 ppm |
Residual solvents | ≤0.1 % (GC) Methanol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...