5-Fluoro-2'-deoxyuridine (floxuridine) 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine
22760-0025
22760-1000
22760-5000
TS22760-0025EA
1266
USD
TS22760-0025
TS22760-1000
TS22760-5000
5-Fluoro-2'-deoxyuridine (floxuridine) 99+%
5-Fluoro-2'-deoxyuridine (floxuridine)
Formula:
C₉H₁₁FN₂O₅ MW: 246.19 g/mol Melting Pt: 148…153 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006530 CAS Number: 50-91-9 EINECS: 200-072-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to almost white Powder |
Infrared spectrum | Conforms |
Melting point | 145 to 153 °C |
Loss on drying | ≤0.5 % (60 °C, 4 hrs) (vacuum) |
Sulfated ash | ≤0.1 % |
HPLC | ≥99.0 % |
Specific optical rotation | +26° to +39° (20 °C, 589 nm) (c=1, Water) |
Heavy metals (as Pb) | ≤20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...