2'-Deoxycytidine 99+%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, deoxyC
22672-0010
22672-0050
22672-0250
22672-2500
22672-5000
TS22672-0010EA
78
USD
TS22672-0010
TS22672-0050
TS22672-0250
TS22672-2500
TS22672-5000
2'-Deoxycytidine 99+%
2'-Deoxycytidine
Formula:
C₉H₁₃N₃O₄ MW: 227.22 g/mol Melting Pt: 209…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006547 CAS Number: 951-77-9 EINECS: 213-454-1 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤8 % |
HPLC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...