2'-Deoxyadenosine monohydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol hydrate
15499-0010
15499-0050
15499-0250
TS15499-0010EA
54
USD
TS15499-0010
TS15499-0050
TS15499-0250
2'-Deoxyadenosine monohydrate 99%
2'-Deoxyadenosine monohydrate
Formula:
C₁₀H₁₃N₅O₃·1H₂O MW: 269.26 g/mol (251.24 anhy) Melting Pt: 185…187 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00149364 CAS Number: 16373-93-6 EINECS: 213-488-7 |
Specification Test Results
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤8 % (K.F.) |
Sulfated ash | ≤0.1 % (800°C) |
HPLC | ≥98.5 % |
Appearance of solution | (c=1, w/v, 1M NaOH) clear colorless to almost colorless |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...