2'-Deoxyguanosine hydrate 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: 9-(2-Deoxy-β-D-ribofuranosyl)guanine hydrate, Guanine-2′-deoxyriboside hydrate, 2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one hydrate(1:x)

22673-0010 22673-0050 22673-1000
TS22673-0010EA 254 USD
TS22673-0010 TS22673-0050 TS22673-1000
2'-Deoxyguanosine hydrate 99+%
2'-Deoxyguanosine hydrate
Formula: C₁₀H₁₃N₅O₄·xH₂O
MW: 267.24 g/mol (anhy)
Melting Pt: 300 °C (dec.)
Flash Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150760
CAS Number: 207121-55-9
EINECS: 213-505-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤8.0 % (K.F.)
HPLC ≥99.0 %
UV spectrophotometry molar absorptivity 13700 +/- 4%, on dry basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR